Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCHS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017639.3 | NP_060109.2 | 13 | 2716 | ||
XM_011532045.1 | XP_011530347.1 | 8 | 4504 | |||
XM_011532046.2 | XP_011530348.1 | 7 | 2160 | |||
GenBank mRNA | AY354497.1 | - | 13 | 2716 |