Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIVM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159596.1 | NP_001153068.1 | 9 | 1325 | ||
NM_017693.3 | NP_060163.2 | 11 | 1988 | |||
GenBank mRNA | AF339783.1 | - | 6 | 579 | ||
AF339793.1 | - | 7 | 1377 | |||
AF411385.1 | - | 11 | 1988 | |||
AK000166.1 | - | 9 | 1200 | |||
AK126285.1 | - | 1 | 1001 | |||
AK225229.1 | - | 9 | 1200 | |||
AK290658.1 | - | 11 | 1857 | |||
AK304391.1 | - | 9 | 1609 | |||
BC020663.1 | - | 9 | 1132 | |||
BC051813.2 | - | 9 | 1113 | |||
BC075084.2 | - | 9 | 1336 | 30915598 | ||
BC075085.2 | - | 9 | 1336 | 30915386 | ||
BC112339.1 | - | 9 | 1080 | 40053977 |
Gene Symbol: BIVM-ERCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204425.1 | NP_001191354.1 | 9 | 1431 | ||
GenBank mRNA | AB209312.1 | - | 9 | 1223 | ||
LM994822.1 | - | 9 | 1309 |