Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYTL2 SIRNA Assays ›
Gene Symbol:
SYTL2
Gene Name:
synaptotagmin like 2
Gene Aliases:
CHR11SYT, EXO4, PPP1R151, SGA72M, SLP2, SLP2A
Chromosome Location:
Chr.11: 85694221 - 85811182 on Build GRCh38
UniGene ID:
Hs.369520
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s29567
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYTL2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001162951.2
NP_001156423.1
18
3043
NM_001162952.2
NP_001156424.1
10
1573
NM_001162953.2
NP_001156425.1
18
3046
NM_001289608.1
NP_001276537.1
19
2852
NM_001289609.1
NP_001276538.1
11
1686
NM_001289610.1
NP_001276539.1
11
1257
NM_032943.4
NP_116561.1
19
3222
NM_206929.3
NP_996812.1
12
1377
NM_206930.3
NP_996813.1
9
1550
XM_005274057.1
XP_005274114.1
18
6682
XM_005274060.1
XP_005274117.1
19
2866
XM_005274061.1
XP_005274118.1
19
2863
XM_005274062.2
XP_005274119.1
20
3287
XM_005274063.1
XP_005274120.1
18
2746
XM_005274066.1
XP_005274123.1
17
2695
XM_005274067.4
XP_005274124.1
11
2989
XM_011545105.1
XP_011543407.1
19
6730
XM_011545106.1
XP_011543408.1
19
6727
XM_011545107.2
XP_011543409.1
20
7151
XM_011545108.1
XP_011543410.1
18
6610
XM_011545109.2
XP_011543411.1
19
6712
XM_011545110.1
XP_011543412.1
18
6559
XM_011545114.2
XP_011543416.1
10
2895
XM_011545115.1
XP_011543417.1
8
989
XM_011545116.2
XP_011543418.1
9
2774
XM_017017933.1
XP_016873422.1
18
6679
XM_017017934.1
XP_016873423.1
20
7148
XM_017017935.1
XP_016873424.1
16
6526
XM_017017936.1
XP_016873425.1
16
6224
XM_017017937.1
XP_016873426.1
19
2767
XM_017017938.1
XP_016873427.1
18
2725
XM_017017939.1
XP_016873428.1
12
3033
XM_017017940.1
XP_016873429.1
11
2985
GenBank mRNA
AB046817.1
-
19
3233
AK024872.1
-
9
1714
AK074737.1
-
12
1334
AK092121.1
-
9
1368
AK124754.1
-
12
2958
AK127504.1
-
13
3920
AK131365.1
-
12
1850
AK222707.1
-
9
1714
AK222710.1
-
9
1714
AK296224.1
-
19
2834
AK296740.1
-
16
2782
AK298604.1
-
11
1175
AL834422.1
-
6
612
AY386362.1
-
12
5237
BC015540.2
-
9
1044
BC068495.1
-
11
1409
BC110315.1
-
10
1102
BC136450.1
-
18
2978
9020439
BC144114.1
-
18
2978
9052634
BC144115.1
-
18
2975
9052635
BX647671.1
-
13
3673
BX648068.1
-
12
3119
EU035829.1
-
18
2699
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
exocytosis
vesicle docking involved in exocytosis
vesicle fusion
negative regulation of phosphatase activity
vesicle-mediated transport
regulation of calcium ion-dependent exocytosis
calcium ion-regulated exocytosis of neurotransmitter
positive regulation of mucus secretion
protein localization to plasma membrane
phosphatidylserine binding
calcium ion binding
protein binding
calcium-dependent phospholipid binding
phosphatidylinositol-4,5-bisphosphate binding
Rab GTPase binding
phosphatase binding
syntaxin binding
clathrin binding
neurexin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0