Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CEACAM4 SIRNA Assays ›
Gene Symbol:
CEACAM4
Gene Name:
carcinoembryonic antigen related cell adhesion molecule 4
Gene Aliases:
CGM7, CGM7_HUMAN, NCA
Chromosome Location:
Chr.19: 41616986 - 41627087 on Build GRCh38
UniGene ID:
Hs.12
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s2959
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CEACAM4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001817.3
NP_001808.2
2
214
XM_006722977.3
XP_006723040.1
2
227
XM_011526343.1
XP_011524645.1
2
214
XM_011526345.2
XP_011524647.1
2
227
XM_011526348.2
XP_011524650.1
2
214
XM_011526349.2
XP_011524651.1
2
227
XM_011526350.2
XP_011524652.1
2
214
XM_011526352.1
XP_011524654.1
2
214
XM_011526354.1
XP_011524656.1
2
214
XM_017026199.1
XP_016881688.1
2
214
XM_017026200.1
XP_016881689.1
2
214
XM_017026201.1
XP_016881690.1
2
227
XM_017026202.1
XP_016881691.1
2
227
XM_017026203.1
XP_016881692.1
2
227
XM_017026204.1
XP_016881693.1
2
227
XM_017026205.1
XP_016881694.1
2
214
XM_017026206.1
XP_016881695.1
2
214
XM_017026207.1
XP_016881696.1
2
227
XM_017026208.1
XP_016881697.1
2
227
XM_017026209.1
XP_016881698.1
2
214
XM_017026210.1
XP_016881699.1
2
214
XM_017026211.1
XP_016881700.1
2
214
XM_017026212.1
XP_016881701.1
2
214
GenBank mRNA
AB924369.1
-
2
103
AB924370.1
-
2
103
AK298701.1
-
2
214
BX644557.1
-
2
215
BX644579.1
-
2
216
D90276.1
-
2
200
Back To Top
More Information
Gene Ontology Categories:
Function(s)
phagocytosis
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0