Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164470.1 | NP_001157942.1 | 2 | 168 | ||
NM_017716.2 | NP_060186.2 | 2 | 168 | |||
XM_011545117.2 | XP_011543419.1 | 3 | 316 | |||
GenBank mRNA | AK000224.1 | - | 2 | 168 | ||
AK309275.1 | - | 3 | 281 | |||
BC029793.1 | - | 2 | 156 | 5185424 | ||
BP262076.1 | - | 2 | 170 | |||
BP262976.1 | - | 2 | 168 |