Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271420.1 | NP_001258349.1 | 8 | 1387 | ||
NM_017775.3 | NP_060245.3 | 8 | 1250 | |||
XM_017024801.1 | XP_016880290.1 | 8 | 1163 | |||
XM_017024802.1 | XP_016880291.1 | 8 | 1163 | |||
GenBank mRNA | AK000350.1 | - | 6 | 466 | ||
AK025958.1 | - | 2 | 136 | |||
AK055780.1 | - | 8 | 943 | |||
AK056878.1 | - | 8 | 1162 | |||
AK094819.1 | - | 1 | 1444 | |||
AK297783.1 | - | 8 | 806 | |||
AL832412.1 | - | 1 | 504 | |||
BC011698.2 | - | 8 | 789 | |||
BC073796.1 | - | 8 | 793 | |||
BC105128.1 | - | 8 | 1250 | 8144131 | ||
BC112107.1 | - | 8 | 1250 | 8327575 |