Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166269.1 | NP_001159741.1 | 5 | 607 | ||
NM_017815.2 | NP_060285.2 | 5 | 599 | |||
GenBank mRNA | AK000431.1 | - | 5 | 568 | ||
AK026016.1 | - | 5 | 553 | |||
BC001916.1 | - | 5 | 596 | 2905474 | ||
BC002554.2 | - | 5 | 530 | 3139198 | ||
BQ066232.1 | - | 5 | 607 | |||
BX247995.1 | - | 4 | 416 |