Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017831.3 | NP_060301.2 | 5 | 908 | ||
XM_011526045.2 | XP_011524347.1 | 5 | 1177 | |||
GenBank mRNA | AK000463.1 | - | 5 | 908 | ||
AK225518.1 | - | 5 | 908 | |||
BC012021.1 | - | 5 | 539 | 4521247 | ||
BU500546.1 | - | 5 | 398 |