Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UCKL1 SIRNA Assays ›
Gene Symbol:
UCKL1
Gene Name:
uridine-cytidine kinase 1 like 1
Gene Aliases:
UCK1L, URKL1
Chromosome Location:
Chr.20: 63939829 - 63959435 on Build GRCh38
UniGene ID:
Hs.504998
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s29867
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UCKL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001193379.1
NP_001180308.1
6
1070
NM_017859.3
NP_060329.2
6
842
NR_126526.1
6
842
XM_005260216.2
XP_005260273.1
6
810
XM_006723806.2
XP_006723869.1
6
810
XM_006723807.2
XP_006723870.1
6
810
XM_006723808.1
XP_006723871.1
6
862
XM_006723809.1
XP_006723872.1
6
842
XM_006723810.1
XP_006723873.1
6
839
XM_006723811.3
XP_006723874.1
6
2831
XM_011528868.2
XP_011527170.1
6
2834
XM_011528869.1
XP_011527171.1
7
4105
XM_011528870.1
XP_011527172.1
6
824
XM_011528871.1
XP_011527173.1
6
851
XM_011528872.1
XP_011527174.1
7
4094
XM_011528873.2
XP_011527175.1
6
810
XM_017027894.1
XP_016883383.1
6
810
XM_017027895.1
XP_016883384.1
6
810
XM_017027896.1
XP_016883385.1
6
842
XM_017027897.1
XP_016883386.1
6
842
XM_017027898.1
XP_016883387.1
6
839
XM_017027899.1
XP_016883388.1
6
839
XM_017027900.1
XP_016883389.1
6
1688
XM_017027901.1
XP_016883390.1
6
1686
XM_017027902.1
XP_016883391.1
7
1042
XM_017027903.1
XP_016883392.1
6
851
XM_017027904.1
XP_016883393.1
6
3766
XM_017027905.1
XP_016883394.1
6
3769
XM_017027906.1
XP_016883395.1
6
3766
XM_017027907.1
XP_016883396.1
2
562
XM_017027908.1
XP_016883397.1
3
465
GenBank mRNA
AJ605558.1
-
6
811
AK000524.1
-
6
811
AK222559.1
-
6
813
AK223220.1
-
6
811
AK301602.1
-
6
750
AK303688.1
-
6
1022
BC033078.1
-
6
780
4810505
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001109212, XM_006235797, XM_006235796, XM_006235795, XM_006235794, and others.
Panther Classification:
Molecular Function -
nucleotide kinase
Gene Ontology Categories:
Function(s)
Process(es)
pyrimidine nucleobase metabolic process
viral process
phosphorylation
pyrimidine nucleoside salvage
UMP salvage
CTP salvage
uridine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-jmz56:80/10.239.163.191:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0