Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017882.2 | NP_060352.1 | 7 | 936 | ||
GenBank mRNA | AK000568.1 | - | 7 | 881 | ||
AK027604.1 | - | 7 | 904 | |||
AK074753.1 | - | 4 | 430 | |||
AK291175.1 | - | 7 | 863 | |||
AK293155.1 | - | 5 | 580 | |||
AK293197.1 | - | 7 | 1112 | |||
BC010849.1 | - | 7 | 850 | 3878776 | ||
BC013130.1 | - | 7 | 787 | 4517748 | ||
CR457244.1 | - | 7 | 778 | |||
KU178671.1 | - | 7 | 778 | |||
KU178672.1 | - | 7 | 897 | |||
KU178673.1 | - | 6 | 679 | |||
KU178674.1 | - | 6 | 589 |