Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165136.1 | NP_001158608.1 | 21 | 2831 | ||
NM_017912.3 | NP_060382.3 | 22 | 2939 | |||
XM_005263083.3 | XP_005263140.1 | 22 | 3136 | |||
XM_011532053.2 | XP_011530355.1 | 21 | 3030 | |||
XM_017008333.1 | XP_016863822.1 | 21 | 2497 | |||
GenBank mRNA | AF336798.1 | - | 22 | 2939 | ||
AK000644.1 | - | 9 | 1027 | |||
AK097168.1 | - | 16 | 1754 | |||
AK225506.1 | - | 9 | 1027 | |||
AK295832.1 | - | 22 | 2849 | |||
AL833664.1 | - | 11 | 1267 | |||
AY653201.1 | - | 21 | 2831 | |||
AY653202.1 | - | 21 | 2743 | |||
AY653203.1 | - | 19 | 2524 | |||
BC035775.1 | - | 8 | 879 | |||
BC042047.1 | - | 22 | 2815 | 5531519 | ||
BX647121.1 | - | 22 | 2615 |