Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017919.2 | NP_060389.2 | 4, 5 | 549 | ||
XM_011518820.2 | XP_011517122.1 | 4, 5 | 632 | |||
XM_011518821.2 | XP_011517123.1 | 4, 5 | 880 | |||
XM_011518823.2 | XP_011517125.1 | 2, 3 | 370 | |||
XM_017014875.1 | XP_016870364.1 | 5, 6 | 726 | |||
GenBank mRNA | AK000658.1 | - | 4, 5 | 498 | ||
AK095262.1 | - | 4, 5 | 1160 | |||
AL833620.1 | - | 4, 5 | 1061 | |||
BC070269.1 | - | 4, 5 | 492 | |||
BC101564.1 | - | 3, 4 | 486 | 8069070 | ||
BC101566.1 | - | 3, 4 | 486 | 8069072 | ||
BC111009.1 | - | 4, 5 | 509 | 5768578 |