Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SREBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004599.3 | NP_004590.2 | 5 | 1201 | ||
NR_103834.1 | 5 | 1201 | ||||
XM_006724310.2 | XP_006724373.1 | 5 | 1445 | |||
XM_011530347.1 | XP_011528649.1 | 5 | 1030 | |||
XM_017028921.1 | XP_016884410.1 | 5 | 1030 | |||
XM_017028922.1 | XP_016884411.1 | 5 | 1030 | |||
GenBank mRNA | AK299914.1 | - | 5 | 1030 | ||
BC051385.1 | - | 5 | 1107 | 5497929 | ||
BC051799.1 | - | 5 | 1167 | |||
BC056158.1 | - | 5 | 1147 | 6169568 | ||
CT841522.1 | - | 5 | 1091 | |||
U02031.1 | - | 5 | 1126 |