Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166373.1 | NP_001159845.1 | 7 | 726 | ||
NM_017923.3 | NP_060393.1 | 3 | 613 | |||
XM_011532055.2 | XP_011530357.1 | 7 | 1431 | |||
XM_017008334.1 | XP_016863823.1 | 6 | 653 | |||
XM_017008335.1 | XP_016863824.1 | 7 | 1032 | |||
XM_017008336.1 | XP_016863825.1 | 7 | 898 | |||
GenBank mRNA | AK000675.1 | - | 3 | 502 | ||
AK289711.1 | - | 7 | 726 | |||
AK301408.1 | - | 4 | 627 | |||
AL713759.1 | - | 7 | 1521 |