Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100818.1 | NP_001094288.1 | 2 | 355 | ||
NM_017933.4 | NP_060403.3 | 3 | 566 | |||
XM_011511384.2 | XP_011509686.1 | 2 | 238 | |||
XM_017004404.1 | XP_016859893.1 | 3 | 5497 | |||
XM_017004405.1 | XP_016859894.1 | 4 | 436 | |||
XM_017004406.1 | XP_016859895.1 | 2 | 94 | |||
GenBank mRNA | AB075874.1 | - | 3 | 221 | ||
AK000708.1 | - | 2 | 194 | |||
AK096636.1 | - | 2 | 297 | |||
AK125359.1 | - | 3 | 490 | |||
AY317148.1 | - | 2 | 187 | |||
DB474424.1 | - | 2 | 355 |