Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017934.5 | NP_060404.3 | 31 | 3835 | ||
XM_005248729.4 | XP_005248786.1 | 31 | 3832 | |||
XM_011535918.2 | XP_011534220.1 | 28 | 3491 | |||
XM_017010989.1 | XP_016866478.1 | 33 | 3977 | |||
XM_017010990.1 | XP_016866479.1 | 30 | 3575 | |||
GenBank mRNA | AF310250.1 | - | 8 | 842 | ||
AJ303102.1 | - | 31 | 3622 | |||
AK000712.1 | - | 4 | 465 | |||
AL161957.1 | - | 9 | 898 | |||
BC008909.2 | - | 6 | 563 | |||
BC064611.1 | - | 3 | 304 | |||
BC081569.1 | - | 9 | 878 | |||
BC137488.1 | - | 31 | 3715 | 9021488 | ||
DQ924532.1 | - | 31 | 3609 |