Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017934.5 | NP_060404.3 | 18 | 2226 | ||
XM_005248729.4 | XP_005248786.1 | 18 | 2226 | |||
XM_011535918.2 | XP_011534220.1 | 15 | 1882 | |||
XM_011535919.1 | XP_011534221.1 | 18 | 2226 | |||
XM_017010989.1 | XP_016866478.1 | 20 | 2368 | |||
XM_017010990.1 | XP_016866479.1 | 17 | 1966 | |||
GenBank mRNA | AJ303102.1 | - | 18 | 2013 | ||
AK057039.1 | - | 18 | 2168 | |||
AK127816.1 | - | 6 | 860 | |||
BC021905.1 | - | 18 | 2115 | |||
BC137488.1 | - | 18 | 2106 | 9021488 | ||
DQ924532.1 | - | 18 | 2000 |