Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHI3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025197.1 | NP_001020368.1 | 4 | 481 | ||
NM_001025199.1 | NP_001020370.1 | 4 | 541 | |||
NM_004000.2 | NP_003991.2 | 5 | 511 | |||
GenBank mRNA | AK223316.1 | - | 4 | 421 | ||
AK298466.1 | - | 4 | 432 | |||
BC011460.1 | - | 5 | 449 | 4214121 | ||
BG698401.1 | - | 5 | 511 | |||
BM787868.1 | - | 4 | 531 | |||
BT006767.1 | - | 5 | 440 | |||
U49835.1 | - | 5 | 460 | |||
U58514.1 | - | 5 | 476 | |||
U58515.1 | - | 4 | 542 |