Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017957.2 | NP_060427.2 | 2 | 488 | ||
GenBank mRNA | AF324241.1 | - | 1 | 53 | ||
AK000785.1 | - | 2 | 437 | |||
AK291658.1 | - | 2 | 437 | |||
AK292977.1 | - | 2 | 437 | |||
AK295730.1 | - | 2 | 296 | |||
AK296346.1 | - | 2 | 437 | |||
BC001038.2 | - | 2 | 417 | |||
BC051365.1 | - | 2 | 417 | |||
BC077722.1 | - | 2 | 417 | |||
BC110602.2 | - | 1 | 183 | |||
BC110605.2 | - | 1 | 183 | 40027860 | ||
DA392550.1 | - | 2 | 451 |