Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZWILCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287821.1 | NP_001274750.1 | 18 | 2054 | ||
NM_001287822.1 | NP_001274751.1 | 17 | 1958 | |||
NM_001287823.1 | NP_001274752.1 | 17 | 1907 | |||
NM_017975.4 | NP_060445.3 | 18 | 1959 | |||
GenBank mRNA | AK023175.1 | - | 18 | 1949 | ||
AK027468.1 | - | 9 | 774 | |||
AK122787.1 | - | 17 | 1746 | |||
AK291048.1 | - | 17 | 1664 | |||
BC036900.1 | - | 18 | 1705 | 5535106 | ||
BC090041.1 | - | 18 | 1803 | 6472990 | ||
BX640701.1 | - | 18 | 2021 |