Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017998.2 | NP_060468.2 | 2 | 1220 | ||
GenBank mRNA | AK000972.1 | - | 2 | 1220 | ||
BC039588.1 | - | 2 | 1154 | 5740394 |