Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198532.1 | NP_001185461.1 | 4 | 466 | ||
NM_001198533.1 | NP_001185462.1 | 5 | 692 | |||
NM_018002.3 | NP_060472.2 | 5 | 692 | |||
NM_181354.4 | NP_851999.2 | 3 | 522 | |||
XM_006716595.2 | XP_006716658.1 | 4 | 461 | |||
XM_017013589.1 | XP_016869078.1 | 3 | 491 | |||
XM_017013590.1 | XP_016869079.1 | 3 | 332 | |||
XM_017013591.1 | XP_016869080.1 | 3 | 422 | |||
XM_017013592.1 | XP_016869081.1 | 3 | 169 | |||
XM_017013593.1 | XP_016869082.1 | 3 | 396 | |||
XM_017013594.1 | XP_016869083.1 | 3 | 396 | |||
XM_017013595.1 | XP_016869084.1 | 3 | 491 | |||
XM_017013596.1 | XP_016869085.1 | 5 | 664 | |||
XM_017013597.1 | XP_016869086.1 | 3 | 325 | |||
XM_017013598.1 | XP_016869087.1 | 3 | 396 | |||
XM_017013599.1 | XP_016869088.1 | 3 | 491 | |||
XM_017013600.1 | XP_016869089.1 | 3 | 348 | |||
GenBank mRNA | AK096148.1 | - | 3 | 202 | ||
AK124441.1 | - | 5 | 625 | |||
AK292899.1 | - | 3 | 474 | |||
AK296561.1 | - | 4 | 466 | |||
AL833193.1 | - | 2 | 103 | |||
BC019691.1 | - | 3 | 339 | |||
BC032710.2 | - | 3 | 464 | 5496492 | ||
BC035484.1 | - | 3 | 476 | |||
FN650108.1 | - | 4 | 502 |