Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198532.1 | NP_001185461.1 | 10 | 2023 | ||
NM_001198533.1 | NP_001185462.1 | 11 | 2249 | |||
NM_018002.3 | NP_060472.2 | 11 | 2249 | |||
NM_181354.4 | NP_851999.2 | 9 | 2079 | |||
XM_006716595.2 | XP_006716658.1 | 10 | 2018 | |||
XM_017013589.1 | XP_016869078.1 | 9 | 2048 | |||
XM_017013590.1 | XP_016869079.1 | 9 | 1889 | |||
XM_017013591.1 | XP_016869080.1 | 9 | 1979 | |||
XM_017013592.1 | XP_016869081.1 | 9 | 1726 | |||
XM_017013593.1 | XP_016869082.1 | 9 | 1953 | |||
XM_017013594.1 | XP_016869083.1 | 9 | 1953 | |||
XM_017013595.1 | XP_016869084.1 | 8 | 1863 | |||
XM_017013596.1 | XP_016869085.1 | 10 | 2036 | |||
XM_017013597.1 | XP_016869086.1 | 8 | 1697 | |||
XM_017013598.1 | XP_016869087.1 | 8 | 1768 | |||
XM_017013599.1 | XP_016869088.1 | 8 | 1863 | |||
XM_017013600.1 | XP_016869089.1 | 8 | 1720 | |||
GenBank mRNA | AF309387.1 | - | 3 | 466 | ||
AK000987.1 | - | 2 | 220 | |||
AK292899.1 | - | 9 | 2031 | |||
AK296561.1 | - | 10 | 2023 | |||
AL833193.1 | - | 8 | 1660 | |||
BC032710.2 | - | 9 | 2021 | 5496492 | ||
BC035484.1 | - | 9 | 2033 | |||
FN650108.1 | - | 10 | 2059 |