Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720187.2 | XP_006720250.1 | 31 | 4951 | ||
XM_017021397.1 | XP_016876886.1 | 31 | 4954 | |||
GenBank mRNA | AK001104.1 | - | 3 | 352 | ||
AK128275.1 | - | 12 | 1399 | |||
AK302641.1 | - | 4 | 575 | |||
BC015016.2 | - | 5 | 682 |