Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300836.1 | NP_001287765.1 | 3, 4 | 343 | ||
NM_018049.2 | NP_060519.1 | 3, 4 | 343 | |||
XM_017026924.1 | XP_016882413.1 | 3, 4 | 666 | |||
XM_017026925.1 | XP_016882414.1 | 3, 4 | 657 | |||
XM_017026926.1 | XP_016882415.1 | 3, 4 | 666 | |||
XM_017026927.1 | XP_016882416.1 | 3, 4 | 657 | |||
XM_017026928.1 | XP_016882417.1 | 3, 4 | 657 | |||
GenBank mRNA | AK001159.1 | - | 3, 4 | 319 | ||
AK097723.1 | - | 4, 5 | 1350 | |||
AK098725.1 | - | 3, 4 | 433 | |||
BC003084.2 | - | 3, 4 | 264 | 2988404 | ||
BC008966.2 | - | 3, 4 | 290 | 2961278 | ||
CN365243.1 | - | 3, 4 | 262 | |||
CR457253.1 | - | 3, 4 | 238 | |||
HY085955.1 | - | 3, 4 | 333 | |||
HY101097.1 | - | 3, 4 | 346 |