Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018079.4 | NP_060549.4 | 21 | 2924 | ||
XM_011532946.1 | XP_011531248.1 | 21 | 2877 | |||
XM_017004418.1 | XP_016859907.1 | 13 | 2478 | |||
GenBank mRNA | AK001241.1 | - | 15 | 1821 | ||
AK055896.1 | - | 6 | 1744 | |||
AK056536.1 | - | 21 | 2894 | |||
AK301122.1 | - | 13 | 1623 | |||
AK316222.1 | - | 13 | 1604 | |||
BC032538.1 | - | 15 | 1899 | 5528417 |