Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM90A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319982.1 | NP_001306911.1 | 6 | 2302 | ||
NM_018088.3 | NP_060558.3 | 7 | 2459 | |||
XM_006719097.3 | XP_006719160.1 | 3 | 2363 | |||
XM_011520719.1 | XP_011519021.1 | 7 | 2450 | |||
XM_017019547.1 | XP_016875036.1 | 7 | 2447 | |||
GenBank mRNA | AK001270.1 | - | 6 | 2288 | ||
BC042608.1 | - | 7 | 2438 | 5167203 |