Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042410.1 | NP_001035869.1 | 10 | 1384 | ||
NM_001282792.1 | NP_001269721.1 | 7 | 1063 | |||
NM_018089.2 | NP_060559.2 | 10 | 1457 | |||
XM_005246663.2 | XP_005246720.1 | 10 | 1376 | |||
XM_011511392.2 | XP_011509694.1 | 10 | 1454 | |||
XM_017004420.1 | XP_016859909.1 | 10 | 1373 | |||
XM_017004421.1 | XP_016859910.1 | 10 | 1457 | |||
GenBank mRNA | AF364318.1 | - | 9 | 1847 | ||
AK001277.1 | - | 10 | 1393 | |||
AK023206.1 | - | 10 | 1384 | |||
AK056060.1 | - | 1 | 608 | |||
AK095304.1 | - | 9 | 1858 | |||
AK225275.1 | - | 10 | 1382 | |||
AK225309.1 | - | 10 | 1358 | |||
BC000238.1 | - | 10 | 1339 | 3351742 | ||
BC008948.2 | - | 10 | 1368 | 2959748 | ||
BC028334.1 | - | 7 | 985 | |||
DQ412564.1 | - | 10 | 1393 |