Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018124.3 | NP_060594.3 | 13 | 2357 | ||
XM_005256021.4 | XP_005256078.1 | 14 | 2490 | |||
XM_005256022.4 | XP_005256079.1 | 13 | 3240 | |||
XM_006721228.3 | XP_006721291.1 | 13 | 2313 | |||
XM_011523191.2 | XP_011521493.1 | 13 | 2310 | |||
XM_017023391.1 | XP_016878880.1 | 14 | 2406 | |||
XM_017023392.1 | XP_016878881.1 | 12 | 2177 | |||
GenBank mRNA | AK001382.1 | - | 8 | 1259 | ||
AK022673.1 | - | 3 | 396 | |||
AK291200.1 | - | 14 | 2457 | |||
AK296727.1 | - | 5 | 850 | |||
AK315786.1 | - | 10 | 1501 | |||
BC002574.2 | - | 11 | 1734 | |||
BC059371.1 | - | 13 | 2306 | 5266451 |