Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGEF10L SIRNA Assays ›
Gene Symbol:
ARHGEF10L
Gene Name:
Rho guanine nucleotide exchange factor 10 like
Gene Aliases:
GrinchGEF
Chromosome Location:
Chr.1: 17520197 - 17697875 on Build GRCh38
UniGene ID:
Hs.443460
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30315
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGEF10L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001011722.2
NP_001011722.2
19
2171
NM_001319837.1
NP_001306766.1
18
2156
NM_001319838.1
NP_001306767.1
10
1395
NM_001328124.1
NP_001315053.1
13
1620
NM_018125.3
NP_060595.3
21
2404
NR_137287.1
22
3151
NR_137288.1
21
3031
XM_005245923.2
XP_005245980.2
20
2263
XM_005245925.2
XP_005245982.2
20
2251
XM_005245927.2
XP_005245984.2
13
1620
XM_005245929.2
XP_005245986.2
12
1605
XM_006710728.1
XP_006710791.1
20
2445
XM_006710729.1
XP_006710792.1
20
2260
XM_006710731.1
XP_006710794.1
20
2442
XM_011541691.2
XP_011539993.1
19
2260
XM_011541692.2
XP_011539994.1
18
2254
XM_011541693.2
XP_011539995.1
18
2245
XM_017001617.1
XP_016857106.1
19
2236
XM_017001618.1
XP_016857107.1
14
1538
XM_017001619.1
XP_016857108.1
19
2269
XM_017001620.1
XP_016857109.1
18
2254
XM_017001621.1
XP_016857110.1
20
2387
XM_017001622.1
XP_016857111.1
19
2372
GenBank mRNA
AB046846.1
-
21
2404
AK001383.1
-
7
862
AK092020.1
-
10
1392
AK126877.1
-
12
2510
AK300264.1
-
12
1568
AK311076.1
-
20
2214
BC065561.1
-
7
906
BC080596.1
-
10
1067
BC117171.1
-
19
2161
40125722
BC143347.1
-
19
2161
9051852
DQ139833.1
-
18
2113
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein modulator
protein-binding activity modulator
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of Rho protein signal transduction
positive regulation of GTPase activity
Rho guanyl-nucleotide exchange factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0