Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PBRM1 SIRNA Assays ›
Gene Symbol:
PBRM1
Gene Name:
polybromo 1
Gene Aliases:
BAF180, PB1
Chromosome Location:
Chr.3: 52545352 - 52685933 on Build GRCh38
UniGene ID:
Hs.189920
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30401
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PBRM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_018313.4
NP_060783.3
17, 18
2673
XM_005265279.4
XP_005265336.1
19, 20
2946
XM_005265280.3
XP_005265337.1
18, 19
2763
XM_005265282.3
XP_005265339.1
19, 20
2905
XM_005265283.3
XP_005265340.1
18, 19
2760
XM_011533900.2
XP_011532202.1
21, 22
3112
XM_011533902.2
XP_011532204.1
20, 21
3044
XM_011533903.2
XP_011532205.1
20, 21
3049
XM_017006725.1
XP_016862214.1
20, 21
3019
XM_017006726.1
XP_016862215.1
21, 22
3129
XM_017006727.1
XP_016862216.1
21, 22
3074
XM_017006728.1
XP_016862217.1
20, 21
3070
XM_017006729.1
XP_016862218.1
20, 21
3478
XM_017006730.1
XP_016862219.1
21, 22
3065
XM_017006731.1
XP_016862220.1
20, 21
3048
XM_017006732.1
XP_016862221.1
20, 21
3044
XM_017006733.1
XP_016862222.1
20, 21
3438
XM_017006734.1
XP_016862223.1
20, 21
3011
XM_017006735.1
XP_016862224.1
19, 20
3432
XM_017006736.1
XP_016862225.1
19, 20
3433
XM_017006737.1
XP_016862226.1
19, 20
3352
XM_017006738.1
XP_016862227.1
19, 20
2973
XM_017006739.1
XP_016862228.1
19, 20
3349
XM_017006740.1
XP_016862229.1
19, 20
2890
XM_017006741.1
XP_016862230.1
19, 20
2981
XM_017006742.1
XP_016862231.1
19, 20
3004
XM_017006743.1
XP_016862232.1
20, 21
3478
XM_017006744.1
XP_016862233.1
19, 20
2970
XM_017006745.1
XP_016862234.1
19, 20
3324
XM_017006746.1
XP_016862235.1
19, 20
3321
XM_017006747.1
XP_016862236.1
19, 20
3383
XM_017006748.1
XP_016862237.1
18, 19
3279
XM_017006749.1
XP_016862238.1
18, 19
2845
XM_017006750.1
XP_016862239.1
17, 18
2733
XM_017006751.1
XP_016862240.1
19, 20
2981
XM_017006752.1
XP_016862241.1
20, 21
3478
XM_017006753.1
XP_016862242.1
20, 21
3477
XM_017006754.1
XP_016862243.1
20, 21
3044
XM_017006755.1
XP_016862244.1
19, 20
2981
XM_017006756.1
XP_016862245.1
19, 20
2981
XM_017006757.1
XP_016862246.1
17, 18
2704
XM_017006758.1
XP_016862247.1
17, 18
2835
XM_017006759.1
XP_016862248.1
19, 20
2981
XM_017006760.1
XP_016862249.1
20, 21
3044
XM_017006761.1
XP_016862250.1
20, 21
3053
XM_017006762.1
XP_016862251.1
20, 21
3044
XM_017006763.1
XP_016862252.1
21, 22
3103
XM_017006764.1
XP_016862253.1
19, 20
2981
XM_017006765.1
XP_016862254.1
18, 19
2860
XM_017006766.1
XP_016862255.1
19, 20
2988
GenBank mRNA
AF177387.1
-
16, 17
2562
AF197569.1
-
17, 18
2655
AF225870.1
-
15, 16
2466
AF225872.1
-
16, 17
2562
AK001926.1
-
1, 2
141
AK056541.1
-
16, 17
2436
AY281068.1
-
16, 17
2561
BC115009.1
-
16, 17
2577
7962003
BC115010.1
-
16, 17
2578
BC115011.1
-
16, 17
2578
7962010
BC129934.1
-
17, 18
2607
40082629
BC129935.1
-
17, 18
2607
40082630
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
negative regulation of cell proliferation
covalent chromatin modification
DNA binding
chromatin binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0