Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P3H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ430351.2 | - | 3 | 890 | ||
AK001580.1 | - | 3 | 281 | |||
AK056447.1 | - | 3 | 890 | |||
BC005029.1 | - | 3 | 275 | 4040181 | ||
DA720646.1 | - | 3 | 447 |