Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193360.1 | NP_001180289.1 | 8 | 1476 | ||
NM_001193361.1 | NP_001180290.1 | 7 | 1392 | |||
NM_001193362.1 | NP_001180291.1 | 8 | 1635 | |||
NM_001193363.1 | NP_001180292.1 | 7 | 1551 | |||
NM_018199.3 | NP_060669.1 | 6 | 1012 | |||
NR_034164.1 | 9 | 1566 | ||||
NR_034165.1 | 9 | 1725 | ||||
XM_005267817.3 | XP_005267874.1 | 6 | 942 | |||
XM_011536908.2 | XP_011535210.1 | 6 | 1030 | |||
XM_017021421.1 | XP_016876910.1 | 7 | 1075 | |||
GenBank mRNA | AK001600.1 | - | 6 | 940 | ||
AK056025.1 | - | 9 | 1512 | |||
AK292563.1 | - | 8 | 1635 | |||
AK295601.1 | - | 7 | 1511 | |||
AL834434.1 | - | 6 | 934 | |||
BC001962.1 | - | 6 | 1072 | 3537432 | ||
BX647767.1 | - | 7 | 1342 | |||
CR936728.1 | - | 8 | 1351 | |||
CR936729.1 | - | 7 | 1359 |