Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018268.3 | NP_060738.2 | 7 | 861 | ||
XM_005248552.3 | XP_005248609.1 | 7 | 828 | |||
XM_011543505.2 | XP_011541807.1 | 7 | 623 | |||
XM_011543507.2 | XP_011541809.1 | 7 | 828 | |||
XM_017009606.1 | XP_016865095.1 | 7 | 1271 | |||
GenBank mRNA | AF115511.1 | - | 7 | 610 | ||
AK001766.1 | - | 7 | 807 | |||
AK021855.1 | - | 7 | 647 | |||
AK023977.1 | - | 7 | 563 | |||
AK296423.1 | - | 6 | 525 | |||
AK300058.1 | - | 5 | 472 | |||
AK304329.1 | - | 7 | 553 | |||
AL834138.1 | - | 1 | 1542 | |||
BC040241.1 | - | 7 | 613 | 3447269 | ||
KU178690.1 | - | 7 | 541 | |||
KU178691.1 | - | 7 | 556 |