Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CASC1 SIRNA Assays ›
Gene Symbol:
CASC1
Gene Name:
cancer susceptibility candidate 1
Gene Aliases:
LAS1, PPP1R54
Chromosome Location:
Chr.12: 25107057 - 25195177 on Build GRCh38
UniGene ID:
Hs.407771
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30578
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CASC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001082972.2
NP_001076441.1
13
1700
NM_001082973.2
NP_001076442.1
12
1492
NM_001204101.2
NP_001191030.1
10
1315
NM_001204102.2
NP_001191031.1
14
1548
NM_001319977.1
NP_001306906.1
11
1400
NM_001319978.1
NP_001306907.1
12
1666
NM_018272.4
NP_060742.3
13
1510
XM_005253406.1
XP_005253463.1
13
1624
XM_005253408.1
XP_005253465.1
12
1386
XM_005253409.1
XP_005253466.1
11
1400
XM_006719101.1
XP_006719164.1
11
1306
XM_011520723.1
XP_011519025.1
13
1624
XM_011520726.1
XP_011519028.1
13
1653
XM_011520727.1
XP_011519029.1
13
1610
XM_017019560.1
XP_016875049.1
12
1589
XM_017019561.1
XP_016875050.1
11
1596
XM_017019562.1
XP_016875051.1
12
1474
XM_017019563.1
XP_016875052.1
11
1188
XM_017019564.1
XP_016875053.1
10
1156
XM_017019565.1
XP_016875054.1
11
1306
XM_017019566.1
XP_016875055.1
11
1517
XM_017019567.1
XP_016875056.1
8
961
XM_017019568.1
XP_016875057.1
9
1100
GenBank mRNA
AK001783.1
-
7
914
AK093102.1
-
13
1477
AK297998.1
-
10
1279
AK301763.1
-
11
1061
AK302235.1
-
12
1442
AK304602.1
-
11
1400
AY423543.1
-
11
1408
BC028415.1
-
13
1641
4823620
BC031047.1
-
13
1491
BC047415.1
-
14
1546
5269578
BC057795.1
-
12
1490
5266518
BC117281.1
-
10
1325
BC117283.1
-
10
1325
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0