Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018279.3 | NP_060749.2 | 1, 2 | 706 | ||
GenBank mRNA | AK001798.1 | - | 1, 2 | 308 | ||
AK223148.1 | - | 1, 2 | 304 | |||
AK315587.1 | - | 1, 2 | 265 | |||
BC008596.1 | - | 1, 2 | 706 | 4154303 |