Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWF19L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303404.1 | NP_001290333.1 | 3, 4 | 279 | ||
NM_001303405.1 | NP_001290334.1 | 3, 4 | 324 | |||
NM_001303407.1 | NP_001290336.1 | 3, 4 | 263 | |||
NM_018294.5 | NP_060764.3 | 3, 4 | 279 | |||
GenBank mRNA | AK001860.1 | - | 3, 4 | 225 | ||
AK097895.1 | - | 3, 4 | 262 | |||
AK225590.1 | - | 3, 4 | 237 | |||
AL832515.1 | - | 3, 4 | 236 | |||
DC299678.1 | - | 3, 4 | 279 | |||
KU178692.1 | - | 3, 4 | 191 | |||
KU178693.1 | - | 3, 4 | 175 |