Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UEVLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040697.2 | NP_001035787.1 | 6 | 693 | ||
NM_001261382.1 | NP_001248311.1 | 5 | 627 | |||
NM_001261383.1 | NP_001248312.1 | 5 | 627 | |||
NM_001261384.1 | NP_001248313.1 | 4 | 444 | |||
NM_001261385.1 | NP_001248314.1 | 6 | 650 | |||
NM_001261386.1 | NP_001248315.1 | 4 | 393 | |||
NM_001297771.1 | NP_001284700.1 | 6 | 693 | |||
NM_018314.4 | NP_060784.3 | 6 | 693 | |||
XM_006718255.2 | XP_006718318.1 | 5 | 522 | |||
XM_011520201.2 | XP_011518503.1 | 2 | 2524 | |||
XM_017017985.1 | XP_016873474.1 | 5 | 620 | |||
XM_017017986.1 | XP_016873475.1 | 5 | 500 | |||
GenBank mRNA | AF503350.1 | - | 6 | 653 | ||
AK001930.1 | - | 6 | 650 | |||
AK295501.1 | - | 4 | 372 | |||
AK296835.1 | - | 5 | 587 | |||
AK301537.1 | - | 5 | 569 | |||
AK314521.1 | - | 6 | 693 | |||
AK316191.1 | - | 5 | 500 | |||
BC011011.1 | - | 6 | 630 | |||
BC064566.1 | - | 5 | 567 | 5576632 | ||
CD109744.1 | - | 4 | 392 |