Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM144 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018342.4 | NP_060812.2 | 6 | 903 | ||
XM_005263110.1 | XP_005263167.1 | 7 | 858 | |||
XM_005263112.3 | XP_005263169.1 | 8 | 893 | |||
XM_006714254.1 | XP_006714317.1 | 7 | 715 | |||
XM_017008366.1 | XP_016863855.1 | 5 | 871 | |||
XM_017008367.1 | XP_016863856.1 | 4 | 443 | |||
XM_017008368.1 | XP_016863857.1 | 5 | 603 | |||
GenBank mRNA | AK002017.1 | - | 5 | 807 | ||
BC026318.1 | - | 6 | 597 | |||
BC036551.1 | - | 5 | 2181 | |||
BC048117.1 | - | 5 | 2176 | |||
BC054487.1 | - | 6 | 651 | 5732836 | ||
BC060875.1 | - | 6 | 629 | 4800219 |