Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP5S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204446.1 | NP_001191375.1 | 3 | 1152 | ||
NM_001204447.1 | NP_001191376.1 | 3 | 1000 | |||
NM_018347.2 | NP_060817.1 | 3 | 1056 | |||
GenBank mRNA | AK002030.1 | - | 3 | 1024 | ||
AK093332.1 | - | 3 | 968 | |||
BC043344.1 | - | 3 | 993 | 5505030 |