Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIS18BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018353.4 | NP_060823.3 | 13 | 3391 | ||
XM_005267833.4 | XP_005267890.1 | 14 | 3438 | |||
XM_011536923.1 | XP_011535225.1 | 10 | 1876 | |||
XM_017021425.1 | XP_016876914.1 | 15 | 3419 | |||
GenBank mRNA | AB067490.1 | - | 13 | 3392 | ||
BC065544.1 | - | 13 | 3182 | 6015439 |