Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018359.3 | NP_060829.2 | 12 | 1480 | ||
NR_028085.1 | 12 | 1455 | ||||
GenBank mRNA | AK002062.1 | - | 12 | 1409 | ||
BC010493.1 | - | 11 | 1376 | 3048997 | ||
BF735169.1 | - | 1 | 62 | |||
CR457278.1 | - | 11 | 1363 |