Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018397.4 | NP_060867.2 | 9 | 1849 | ||
XM_006713250.3 | XP_006713313.1 | 9 | 1631 | |||
XM_006713251.3 | XP_006713314.1 | 9 | 1808 | |||
XM_006713252.3 | XP_006713315.1 | 8 | 1488 | |||
XM_011533938.2 | XP_011532240.1 | 9 | 1665 | |||
XM_011533939.2 | XP_011532241.1 | 9 | 2504 | |||
XM_017006797.1 | XP_016862286.1 | 9 | 1662 | |||
XM_017006798.1 | XP_016862287.1 | 9 | 1654 | |||
XM_017006799.1 | XP_016862288.1 | 8 | 1485 | |||
GenBank mRNA | AJ272267.1 | - | 7 | 1073 | ||
AK055402.1 | - | 1 | 1237 | |||
AK297575.1 | - | 5 | 532 | |||
BC034502.1 | - | 9 | 1607 | 4824572 |