Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276320.1 | NP_001263249.1 | 4 | 743 | ||
NM_018566.3 | NP_061036.3 | 2 | 717 | |||
GenBank mRNA | AK128014.1 | - | 4 | 743 | ||
AL833081.2 | - | 2 | 649 | |||
BC137166.1 | - | 2 | 709 | 9021163 | ||
BC137167.1 | - | 2 | 709 | 9021164 |