Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037639.2 | NP_001032728.1 | 9 | 1011 | ||
NM_001324436.1 | NP_001311365.1 | 9 | 1059 | |||
NM_001324437.1 | NP_001311366.1 | 8 | 909 | |||
NM_001324438.1 | NP_001311367.1 | 8 | 961 | |||
NM_018622.6 | NP_061092.3 | 10 | 1161 | |||
NR_136893.1 | 10 | 1217 | ||||
GenBank mRNA | AF197937.1 | - | 10 | 1099 | ||
AK024765.1 | - | 10 | 1133 | |||
BC003653.1 | - | 9 | 998 | 3537438 | ||
BC014058.2 | - | 10 | 1119 | 3937238 |