Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017514.4 | NP_059984.3 | 2 | 328 | ||
XM_005274705.4 | XP_005274762.2 | 2 | 322 | |||
XM_006724829.3 | XP_006724892.1 | 2 | 322 | |||
GenBank mRNA | CN427844.1 | - | 1 | 56 | ||
HY098587.1 | - | 2 | 330 | |||
HY158513.1 | - | 2 | 329 | |||
X87852.1 | - | 2 | 337 |