Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXRED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017547.3 | NP_060017.1 | 2 | 273 | ||
NR_037648.1 | 2 | 292 | ||||
XM_011542895.2 | XP_011541197.1 | 2 | 370 | |||
XM_011542896.2 | XP_011541198.1 | 2 | 159 | |||
XM_017018000.1 | XP_016873489.1 | 2 | 248 | |||
XM_017018001.1 | XP_016873490.1 | 2 | 372 | |||
XM_017018003.1 | XP_016873492.1 | 2 | 248 | |||
XM_017018004.1 | XP_016873493.1 | 2 | 370 | |||
XM_017018005.1 | XP_016873494.1 | 1 | 1405 | |||
XM_017018006.1 | XP_016873495.1 | 2 | 248 | |||
GenBank mRNA | AF103801.1 | - | 2 | 171 | ||
AF447877.1 | - | 1 | 520 | |||
AK023987.1 | - | 2 | 142 | |||
AK295267.1 | - | 2 | 228 | |||
AK295428.1 | - | 2 | 142 | |||
AL136923.1 | - | 2 | 153 | |||
BC013902.2 | - | 2 | 129 | 3956972 | ||
DA056004.1 | - | 2 | 273 |