Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other VEZT SIRNA Assays ›
Gene Symbol:
VEZT
Gene Name:
vezatin, adherens junctions transmembrane protein
Gene Aliases:
VEZATIN
Chromosome Location:
Chr.12: 95217746 - 95302799 on Build GRCh38
UniGene ID:
Hs.24135
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31032
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: VEZT
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017599.3
NP_060069.3
9
1556
NR_038241.1
10
1629
NR_038242.1
9
1564
XM_006719477.3
XP_006719540.1
9
1568
XM_006719478.2
XP_006719541.1
8
1462
XM_006719479.2
XP_006719542.1
10
1639
XM_006719480.3
XP_006719543.1
10
1616
XM_006719481.2
XP_006719544.1
9
1486
XM_006719482.3
XP_006719545.1
9
1867
XM_006719483.3
XP_006719546.1
9
1884
XM_006719484.3
XP_006719547.1
8
1794
XM_006719485.2
XP_006719548.1
7
1236
XM_006719486.3
XP_006719549.1
7
1224
XM_006719490.3
XP_006719553.1
10
1627
XM_011538543.1
XP_011536845.1
10
1561
XM_011538544.1
XP_011536846.1
10
1549
XM_011538545.2
XP_011536847.1
9
1529
XM_011538546.1
XP_011536848.1
11
1632
XM_011538547.1
XP_011536849.1
11
1676
XM_011538548.2
XP_011536850.1
8
1322
XM_011538549.1
XP_011536851.1
10
1561
XM_011538550.2
XP_011536852.1
10
1561
XM_011538551.2
XP_011536853.1
10
1549
XM_011538552.1
XP_011536854.1
10
1635
XM_011538553.2
XP_011536855.1
10
1565
XM_017019586.1
XP_016875075.1
9
1517
XM_017019587.1
XP_016875076.1
8
1450
XM_017019588.1
XP_016875077.1
11
1620
XM_017019589.1
XP_016875078.1
10
1604
XM_017019590.1
XP_016875079.1
11
1664
XM_017019591.1
XP_016875080.1
9
1855
XM_017019592.1
XP_016875081.1
8
1309
XM_017019593.1
XP_016875082.1
11
1684
XM_017019594.1
XP_016875083.1
10
1546
XM_017019595.1
XP_016875084.1
9
1568
XM_017019596.1
XP_016875085.1
9
1568
XM_017019597.1
XP_016875086.1
9
1556
XM_017019598.1
XP_016875087.1
9
1556
XM_017019599.1
XP_016875088.1
9
1517
XM_017019600.1
XP_016875089.1
9
1872
XM_017019601.1
XP_016875090.1
10
1623
XM_017019602.1
XP_016875091.1
11
1620
XM_017019603.1
XP_016875092.1
9
1884
XM_017019604.1
XP_016875093.1
7
1236
XM_017019605.1
XP_016875094.1
7
1224
GenBank mRNA
AF216644.1
-
9
1483
AF225417.1
-
9
1527
AF277625.1
-
5
995
AK001338.1
-
7
952
AK307649.1
-
10
1635
AK309943.1
-
9
1503
AL133113.1
-
2
282
AL137497.1
-
10
1551
BC028597.1
-
12
1885
BC064939.1
-
10
1546
6159768
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
single organismal cell-cell adhesion
myosin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0