Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017619.3 | NP_060089.1 | 13 | 1514 | ||
GenBank mRNA | AK057799.1 | - | 13 | 1493 | ||
AK289844.1 | - | 13 | 1776 | |||
AL137730.1 | - | 5 | 2505 | |||
AY099329.1 | - | 13 | 1499 | |||
BC010697.1 | - | 13 | 1490 | 3873751 |