Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017631.5 | NP_060101.3 | 36 | 5257 | ||
XM_011532103.2 | XP_011530405.1 | 36 | 5257 | |||
GenBank mRNA | AK000042.1 | - | 8 | 1058 | ||
AK026333.1 | - | 4 | 523 | |||
AK223184.1 | - | 8 | 1058 | |||
AK225191.1 | - | 8 | 1045 | |||
AL137464.1 | - | 3 | 443 | |||
BC020601.1 | - | 4 | 473 | |||
BC038115.2 | - | 36 | 5202 | 5261001 | ||
BC074781.2 | - | 8 | 1036 | |||
BX648655.1 | - | 16 | 2099 |